Boreal Compound Microscopes with 8" LCD Tablet
Supplier: VWR International
Boreal2
D-EL401 D-EL301
470222-468EA 1370.25 USD
470222-468 470222-470
Boreal Compound Microscopes with 8" LCD Tablet
Microscopes
Use it as a traditional microscope, or view and share live images with the tablet screen.
  • LCD Android Tablet, Eight Inches
  • Screen Resolution: 800 x 1280
  • Built-in Motic Hub for Wireless Sharing
  • Motic Images Plus Software Included
These microscopes combine high speed imaging technology with our most popular microscopes.
The EM model is coarse focus only and includes 4×, 10× and 40× objectives and gliding stage with stage clips. The HM model has coarse and fine focus, 4×, 10×, 40× and 100× objectives and a built-in mechanical stage. Both models have LED cordless illumination.
The Wi-Fi digital tablets transmit live images to iOS or Android devices. Use them as conventional microscopes or share live images with colleagues using Wi-Fi tablets or wireless laptops. The tablet includes preloaded Motic apps.
Información para pedidos: This kit includes a lifetime manufacturer's warranty on the microscope chassis and a one-year warranty on the tablet.