Polypropylene Cylinders
Supplier: VWR International
CC2020/4 CC2020/6 CC2020/3 CC2020/2 CC2020/4 CC2020/5 CC2020/6 CC2020/5 28458-6000_SE CC2020/7 CP1000 CC2020/3
470188-958CS 41.85 USD
470188-958 470148-670 470148-671 470177-819 470188-960 470144-832 470148-837 470177-818 470148-836 470177-817 470177-816 470158-130
Polypropylene Cylinders
Cylinders Measuring Cylinders
Unbreakable Polypropylene.
  • Permanent, molded, single scale metric markings
  • Flared top rims and large spouts facilitate pouring
  • Hexagonal shaped bases prevent tipping and rolling
  • Autoclavable at 121°C
Chemically resistant to most acids, bases, alcohols, esters, and ketones.