Burner Wing Top
Supplier: VWR International
H-6650 C-1505/1 H-6650
470148-920CS 85.57 USD
470202-860 470148-920 18026-108
Burner Wing Top
Heaters Gas Burners
It provides a broad, flat flame and fits mixing tubes with 1/2" outer diameter without stabilizer or 11/16" with stabilizer.