Fast Release Pi-Pumps
Supplier: BEL-ART PRODUCTS SE
F37904-0025 F37904-0002 F37904-0010
470145-290CS 367 USD
470145-290 470145-288 470153-806
Fast Release Pi-Pumps
Pipets Pipet Controllers
Designed for fast and accurate one-handed operation, this pump will reduce the risk of injury from repetitive pipetting.
  • Each pump is resistant to acids, bases, and most solvents
  • Disassembly for cleaning is easy
The fast-release trigger expels the entire contents at one shot, or you can use the thumb wheel for precision dispensing.