Individual Glass Lenses
Supplier: United Scientific Supplies
PH0534K-CV LCV315 PH0536J-CV LCV505 LCV520 LCV708 PH0536L-CV PH0534L-V500 LCV304 PH0536A-CC LCV540 PH0536D-CC PH0534A-CV 3309-08 PH0534L-CV PH0534J-CV PH0534I-CV PH0534H-CV PH0534G-CV PH0536C-CC 3305-08 PH0534F-CV LCC708 PH0536B-CC PH0536H-CV PH0534L PH0534C-CV PH0534B-CV PH0536I-CV PH0534D-CV 4-90936 3309-07
470144-836CS 52.3 USD
470144-836 470111-266 470144-834 470111-268 470135-010 470111-262 470229-794 470111-264 470013-320 470325-670 470111-260 470017-098 470014-580 470005-602 470016-914 470111-258 470014-716 470105-726 470111-256 470007-088 470007-086 470111-270 470013-318 470104-326 470017-124 470104-324 470021-842 470104-328 470016-868 470014-576 470135-008 470014-578
Individual Glass Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
All Types and Sizes for all Optical Applications.
  • Variety of sizes and focal lengths
  • Glass material
This comprehensive range has the right lens for your teaching application. All the lenses are carefully ground and polished to close tolerances and have cleanly finished edges. They are made of optical quality glass, and are uncoated for better abrasion resistance and economy.