Cotton-Tipped Swab Applicators, SciEd Rochester Catalog Divisi
Supplier: VWR
CWSB031WR8 WARD470019-172 999736
470019-172EA 9 USD
470019-172 470150-658 470150-426
Cotton-Tipped Swab Applicators, SciEd Rochester Catalog Divisi
Swabs Swabs for Sampling
Sterile swabs have wooden shafts and are indivdually sealed. Depending on the pack size ordered, they may be packaged in a bag (pack/10) or box (pack/100). Excellent for spreading a confluent lawn of bacteria without breaking the agar surface.
Non sterile swabs are packaged together in a box or bag. Excellent for manipulating small samples that do not need to be kept sterile.