Lens Sets
Supplier: VWR International
PH0538C LSTA50 PH0538A PH0538B PH0538D
470014-448EA 25.3 USD
470014-448 470135-014 470135-012 470018-264 470014-446
Lens Sets
Educational Materials Physics Educational Materials Optics Learning Activities
Choose high quality crown glass or shatter-resistant acrylic lenses.
  • Wide range of lens sizes and materials
  • 6 lenses: double convex, plano-convex, concavo-convex, double concave, plano-concave, and convexo-concave
Perfect for optical benches or general use, these sets contain one each of the six lens types shown. Glass lenses resist scratching, but are more brittle than acrylic. Sets come in a cardboard storage box. Acrylic sets include an activity guide. Available in 38 mm or 50 mm diameter.