VWR® Irradiated Coveralls
Supplier: VWR, part of Avantor
VWR®
414004-452 414004-447 414004-451 414004-449 414004-448 414004-450 89136-084 414004-443 414004-445 414004-444 89136-080 414004-446 414004-453 89136-086 89136-082 414004-454
414004-447CS 355.75 USD
414004-447 414004-446 414004-445 414004-444 414004-443 414004-454 414004-453 414004-452 414004-451 414004-449 414004-448 89136-080 89136-082 89136-084 89136-086 414004-450
VWR® Irradiated Coveralls
Disposable Garments
Irradiated coveralls are manufactured from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection.
  • Excellent fluid and particulate barrier
  • Extremely low level of particle shedding
  • Superior breathability and water vapor transmission rate
  • Soft, cloth-like fabric
Garments exhibit excellent water vapor tranmission to optimize user comfort. Coveralls are gamma irradiated to a Sterility Assurance Level (SAL) of 10-4 to address the concern of bio-burden contamination. Manufactured and packaged in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.
Certificaciones: Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Información para pedidos: Packaged individually in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.