VWR® Disposable Transfer Pipets
Supplier: VWR, part of Avantor
VWR®
414004-002 414004-029 16001-174 414004-038 414004-022 414004-032 414004-027 16001-186 414004-008 16001-170 414004-033 414004-026 414004-046 414004-030 16001-192 414004-003 414004-028 16001-178 414004-011 414004-048 414004-031 414004-010 414004-020 16001-190 414004-006 414004-005 414004-023 414004-012 414004-001 414004-009 414004-042 414004-018 16001-196 414004-007 414004-041 414004-019 414004-021 16001-194 414004-039 414004-047 414004-004
414004-018CS 735.39 USD
414004-018 414004-039 414004-038 16001-178 414004-012 414004-033 414004-011 414004-019 16001-190 414004-021 16001-174 414004-020 414004-042 16001-196 414004-041 16001-170 16001-192 16001-194 414004-029 414004-007 414004-028 414004-006 414004-027 414004-005 414004-026 414004-048 414004-004 414004-003 414004-047 414004-002 414004-046 414004-023 414004-001 414004-022 414004-009 414004-008 414004-032 414004-010 414004-031 16001-186 414004-030
VWR® Disposable Transfer Pipets
Pipets Pasteur/Transfer Pipets
These unbreakable all-in-one pipets eliminate the hazard of broken glass and exposure to infectious materials.
  • Will not shatter or break
  • Can be used in liquid nitrogen
  • Nontoxic and inert
  • No bulb to insert or remove
  • Uniform drop size
Molded from see-through low density polyethylene, they are inert to biological fluids and most acids. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Transfer pipets can be sealed and refrigerated, and will work well whenever there is a need for quick, safe fluid transfer. Can be gas sterilized.