VWR® CERTICLEAN® Class 10 Nitrile Gloves
Supplier: VWR, part of Avantor
VWR® CERTICLEAN®
56979 56975 56978 56974 56975 56977 56976 56974 56977 56976 40101-357 56979
40101-352CS 796.25 USD
CA40101-358 CA40101-357 40101-352 40101-353 CA40101-352 40101-354 CA40101-354 CA40101-353 40101-356 CA40101-356 40101-357 40101-358
VWR® CERTICLEAN® Class 10 Nitrile Gloves
Gloves
Ambidextrous, powder-free gloves with beaded cuffs for strength and reduction of cuff roll down.
  • Superior strength and puncture resistance
  • Certificates of analysis available
  • Quality data available
Nitrile is superior in strength and puncture resistance compared to other glove compounds. Film thickness designed for comfort and tactile sensitivity. Gloves are 6.3 mil thick at fingers and 5.1 mil thick at palms. Textured fingertips for a reliable grip. Uncompromising manufacturing processes yield consistently low particle counts. Proprietary nitrile, silicone-free formulation, and specialized manufacturing processes result in consistently low levels of ionic extractables and nonvolatile residue (NVR) transfer. In-use static dissipative properties superior to natural rubber latex.
The 30.5 cm (12") length and tapered cuff provide more coverage over the garment sleeve.
Envase: Packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Packaged in cleanroom-compatible polyethylene, 100 gloves per double poly bag, 10 bags per case.