PIPETBOY acu 2 Pipet Aid, INTEGRA Biosciences
Supplier: Integra Biosciences
155017 155000 155015 155016 155017 155018 155019
37001-858EA 1097.89 USD
37001-858 37001-856 37001-864 37001-862 37001-860 14222-180 MSPP-155017
PIPETBOY acu 2 Pipet Aid, INTEGRA Biosciences
Pipets Pipet Controllers
This second generation model, the PIPETBOY acu 2 Pipet Aid, is equipped with the latest technology to provide increased pipetting speed and a significant weight reduction in the instrument.
These two features help users to cut work time while offering an improved ergonomic pipetting experience. Besides these updates, the main benefits of the PIPETBOY acu Pipet Aid were preserved, including the unmatched pipetting precision, the colorful look, and the weight balance. Lighter and faster, the world’s most popular pipettor just got better.
Información para pedidos: PIPETBOY acu 2 Pipet Aids are supplied complete with Li-Ion battery, mains adapter, spare sterile 0.45 µm filter, and wall mount. A free Swiss pocket tool is included with each new PIPETBOY acu 2.