Nalgene® Pipet Jars, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
5242-0040 5242-0060 5242-0050
36322-122CS 933.94 USD
36322-122 36322-100 36322-086
Nalgene® Pipet Jars, Thermo Scientific
Pipets Pipet Washers
Cross-linked high-density polyethylene pipet jars are ideal for soaking and washing pipets and glassware, and as large general-purpose containers
Caution: Concentrated sulfuric acid will cause jar to fail. When using sulfuric acid dichromate compound, the acid concentration must be 60% or less. See label on jar. Do not mix sulfuric acid dichromate compound in jar. Keep jar on drain board, in sink, or in container large enough to contain contents in case of leakage.
Tough, stiff, seamless jars resist stress-cracking. All sizes have flanged base for added stability. Breakage is sharply reduced and accidents avoided.
Información para pedidos: Supplied with dust covers.