Standard Wide Mouth Bottles, Vacuum and Ionized, Clear, Qorpak®
Supplier: Qorpak
GLC-01759 GLC-01746 GLC-01803 GLC-01790 GLC-01756 GLC-01817 GLC-01787 GLC-01885 GLC-01861 GLC-01780 GLC-06457 GLC-01836 GLC-01869 GLC-01773
36319-794CS 131.14 USD
36319-794 36319-793 11311-402 36319-790 TN7468C 36319-792 11311-478 36319-791 76487-370 89204-698 76487-374 76487-372 89204-696 36319-744
Standard Wide Mouth Bottles, Vacuum and Ionized, Clear, Qorpak®
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with the product in any way.