VWR® Soil Analysis ASTM Hydrometers, Traceable to NIST
Supplier: VWR, part of Avantor
VWR®
61891-3111 61894-0111
34793-010CS 1448.11 USD
34793-010 34792-001
VWR® Soil Analysis ASTM Hydrometers, Traceable to NIST
Hydrometers
Used to determine the specific gravity of soil in the range of 0.995 to 1.038, or -5 to 60 g/L (using a weight by volume scale), these hydrometers conform to the accuracy requirements of ASTM hydrometers.
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 68 /68 °F
  • Hand inspected by our U.S. quality team
Certificaciones: Traceable to NIST.
Envase: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Precaución: Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.