VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST
Supplier: VWR, part of Avantor
VWR®
61801-0711 60171-4511 61801-0011 61801-0111 61801-2111 61801-1511 61801-0511 61801-1411 61801-2011 60171-4411 61801-1011 61801-1711 61801-1111 61801-0311 61801-1811 61801-0811 61801-0211 61801-1611 61801-1311 61801-0911 61801-1211 61801-0411 61801-0611
34627-650EA 64.71 USD
34627-650 34627-560 34627-231 34627-253 34627-297 34627-173 34627-130 34627-174 34627-050 34627-082 10186-290 10186-288 34627-628 34627-537 34627-504 34627-118 34627-027 34627-479 34627-446 34627-413 34627-311 34627-355 34627-388
VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST
Hydrometers
Available in a wide range of models measuring specific gravity/relative density (g/cmᶟ) of a variety of liquids in various ranges from 0.600 to 1.920, with 0.0005 divisions.
  • 325 mm (12.8”) long
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
  • Hand inspected by our U.S. quality team
Specific gravity hydrometers are the most widely used hydrometer, common in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Envase: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.