VWR® Powder Funnels, Parallel Stems, Polypropylene
Supplier: VWR, part of Avantor
VWR®
400700-0080 400700-0100 400700-0150 400700-0065
30252-953CS 334.24 USD
30252-953 30252-955 30252-957 30252-951
VWR® Powder Funnels, Parallel Stems, Polypropylene
Funnels Powder Funnels
Powder funnels are fast and efficient.
  • Autoclavable
  • Resistant to most chemicals
  • Polypropylene construction
Parallel stems reduce the bridging of powders; external ribbing prevents airlock. Funnels resist most chemicals except strong oxidizing agents. May be used from 0 to 135°C (32 to 275°F).