VWR® Grade 415 Filter Paper, Qualitative, Crepe
Supplier: VWR, part of Avantor
VWR®
28320-019 28320-223 28320-020 28320-121 28320-187 28297-846 28297-846 28320-085 28320-041 28320-100 28320-143 28320-223 28320-121 28320-201
28320-019PK 12.76 USD
28320-019 28320-223 28320-020 28320-121 28320-187 CA28297-846 28297-846 28320-085 28320-041 28320-100 28320-143 CA28320-223 CA28320-121 28320-201
VWR® Grade 415 Filter Paper, Qualitative, Crepe
Filters Filter Paper Qualitative Filter Paper
Good for coarse precipitates such as aluminum hydroxide, cobalt carbonate, mercury iodide, magnesium hydroxide, and more.
  • Coarse porosity
  • Fast flow rate
  • Assorted sizes