VWR® Membrane Air Dryers
Supplier: VWR, part of Avantor
VWR®
64-02-L1466 64-01-L1466
26000-032EA 4949.25 USD
26000-032 26000-030
VWR® Membrane Air Dryers
Heaters Hot Air Dryers
Ideal for supplying compressed air to NMR spectrometers, AA spectrometers, electron microscopes, and thermal analyzers.
  • No electricity required
  • No refrigerants
  • Explosion-proof
Dryers supply oil- and particulate-free dry compressed air at dew points as low as -40°C (-40°F) and at flow rates up to 25scfm. They use the highest efficiency membrane available for low cost operation and minimal maintenance. The dryers can be easily installed on an existing air line, making them ideal for remote and point-of-use installations or for hazardous areas.