Graduated Cylinders, PMP, Kartell
Supplier: Kartell
VWR®
K1575-VWR K1573-VWR K1574-VWR K1577-VWR K1572-VWR K1576-VWR K1571-VWR K1570-VWR
24774-750CS 272.94 USD
24774-750 24774-692 24774-728 24774-785 24774-660 24774-783 24774-637 24774-604
Graduated Cylinders, PMP, Kartell
Cylinders Measuring Cylinders
These polymethylpentene cylinders are transparent, shatterproof, and autoclavable.
  • Transparent
  • Shatterproof
  • Autoclavable
  • Polymethylpentene fabrication
Graduated cylinders withstand temperatures of 0 to 120°C (32 to 248°F) when used continuously, and withstand temperatures as high as 170°C (338°F) when used for short periods of time. Permanent graduations molded in. Meet stringent international quality criteria according to ISO 6706 certification. Not resistant to strong oxidizers, some hydrocarbons, and chlorinated solvents.