VWR® Corks
Supplier: VWR, part of Avantor
VWR®
53924 50224 54518 54125 52424 52224 52024 50824 54525 50725 53925 53925 50724 54524 50924 50324 50924 50824 50024 52824 50424 51624 54527 50625 50625 50825 52224 53724 51224 51724 51224 50225 50524 51024 50124 50325 52824 51324 51724 54523 51524 54523 52024 54526 52424 51624 50424 50025 51524 50624 50025 50825 54526 50325 54522 50224 50425 51824 50525 50725 51124 54125 50124 50125 50425 51824
23420-050PK 44.87 USD
23420-050 23420-490 23420-093 CA23420-388 CA23420-300 23420-413 CA23420-107 CA23420-548 CA23420-468 23420-173 CA23420-140 CA23420-060 23420-130 CA23420-220 23420-210 CA23420-023 23420-027 CA23420-038 CA23420-399 23420-504 23420-548 CA23420-559 CA23420-515 23420-107 CA23420-479 CA23420-435 23420-140 23420-184 CA23420-195 CA23420-151 CA23420-071 CA23420-231 23420-220 23420-264 23420-300 23420-071 23420-038 23420-479 CA23420-366 23420-435 23420-515 23420-559 23420-118 23420-195 CA23420-480 23420-151 CA23420-082 CA23420-242 23420-231 23420-399 CA23420-560 23420-082 23420-526 23420-129 23420-209 CA23420-413 23420-162 23420-480 CA23420-490 CA23420-050 CA23420-210 23420-242 23420-286 23420-322 23420-366 CA23420-130
VWR® Corks
Stoppers Cork Stoppers
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
  • Regular length taper
  • Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade
Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.
This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.