VWR® Wafer Box Sealing Tape, Polyethylene
Supplier: VWR, part of Avantor
VWR®
52B-2DG 52B-34OR 52B-2TR 52B-1TRSMF 52B-2WH 52B-1DB 52B-2OR 52B-1WH 52B-1BL 52B-12RE 52B-1RE 52B-1OR 52B-2DB 52B-2RE 52B-1YE 52B-1DG 52B-3WH 52B-1TR 52B-34TR 52B-2YE
89402-996CS 919.77 USD
89402-996 22221-171 11211-928 89501-298 11211-926 11211-924 89133-258 22221-138 11211-934 11211-922 11211-910 22221-169 11211-932 11211-920 11211-930 22221-163 22221-161 22221-112 43300-976 22221-165
VWR® Wafer Box Sealing Tape, Polyethylene
Tapes
Ideal for packaging and transporting of wafer boxes, this low-density polyethylene tape is low in halogens and leachable chlorides.
  • Metal free adhesive
  • Polyethylene film fabrication
  • Acrylic adhesive minimizes outgassing
  • Excellent chemical resistance
  • Good conformability
Polyethylene film and acrylic adhesive minimize outgassing and offer good 'clean peel' properties. Offers excellent chemical resistance and good conformability.