Motic SMZ-140 Stereo Microscope
Supplier: Motic
1100200600513 1100200600513 1100200600421 1100200600543 1100200600421 1100200600161 1100200600543 1100200600101 1100200600161 1100200600101
19000-408EA 798.04 USD
19000-408 CA19000-408 CA19000-864 CA19000-410 19000-864 CA19000-862 19000-410 CA19000-404 19000-862 19000-404
Motic SMZ-140 Stereo Microscope
Microscopes
The SMZ-140 line provides huge versatility in a stereo microscope system
The SMZ-140 comes with Widefield eyepieces (10x/F.N. 20) with a 4 : 1 zoom ratio, giving a magnification range of 1x to 4x, and a working distance of 80mm. The main power supply is rated at 110V.
With a Greenough Zoom Optical System, the SMZ-140 series allows users to focus at low (10x) magnification and zoom continuously up to 40x for focus on fine details. The SMZ-140 line is available with an array of stand and illumination options for optimization of transmitted and/or incident illumination and working space.
Información de suministro: Microscope is supplied with all accessories listed above. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.