BRAND Reagant Reservoir, BrandTech
Supplier: Brand
703411 703459 703409
80092-454PK 574.5 USD
80092-454 18900-320 80092-456
BRAND Reagant Reservoir, BrandTech
Reagent Reservoirs
This reagent reservoir is made from heavy-duty polypropylene to withstand autoclaving.
  • Made of rugged, reusable polypropylene
  • Autoclavable at 121 °C for 20 minutes
  • Supplied with lid
A lid is included to help protect against sample contamination. Suitable for use with all multichannel pipettes.