BRAND Transferpettor® Single-Channel Positive Displacement Pipettors, Variable Volume, BrandTech
Supplier: BrandTech
Transferpettor™
702810 702812 702804 702806
18900-086EA 724.56 USD
18900-086 18900-088 18900-090 18900-092
BRAND Transferpettor® Single-Channel Positive Displacement Pipettors, Variable Volume, BrandTech
Pipets Micropipets
These pipettors are the solution for many liquids that cannot be pipetted accurately with an air displacement pipet, such as viscous, volatile, or foaming liquids.
  • Instruments, tips, and seals are not autoclavable
  • Optional Transferpettor station holds two instruments
  • Tips and seals for convenient access in the workplace
Pipettors are useful when working with liquids such as concentrated protein solutions, oils, fats, resins, and glycerin. They can be used to accurately pipet surfactants and are designed for use with high-vapor pressure media, such as alcohol, acetone, and hydrocarbons.
The tightly fitting piston comes in direct contact with liquid for reproducible results, without calibration adjustments. Tip walls are wiped clean to the last drop, for repeated use of the tips without cross-contamination.
Información para pedidos: Supplied with manufacturer’s one-year warranty. Pipettors are supplied with one tip and seal, seal mounting/removal tool, performance certificate, and operating manual.