SP Bel-Art Check Valve, Polyethylene, Bel-Art Products, a part of SP
Supplier: Bel-Art
F19713-0002 F19713-0001
16126-558DZ 105.06 USD
16126-558 16126-556
SP Bel-Art Check Valve, Polyethylene, Bel-Art Products, a part of SP
Valves Vacuum Valves
Ensure one-way directional flow in vacuum lines.
  • Polyethylene construction
  • Neoprene disk without metal
  • Built-in arrows indicate free flow direction
  • Positive operation
These polyethylene plastic check valves operate positively and will instantly block flow if direction changes. Built-in arrows indicate direction of free flow. Neoprene disk, no metal. Maximum operating pressure is 20psi.