Prefilled Formalin Containers, Azer Scientific

Supplier: Azer Scientific
PFNBF-3840 PFNBF-1.25G PFNBF-0.6G B-PFNBF-40 B-PFNBF120 PFNBF-0.6G B-PFNBF-90 B-PFNBF-60 PFNBF-1.25G B-PFNBF-20
89132-039CS 547.06 USD
CA10014-686 89132-039 CA10014-362 89132-041 10014-686 89132-043 95044-215 89132-045 10014-362 16008-002
Prefilled Formalin Containers, Azer Scientific
Sample Containers General Purpose Containers
Prefilled formalin containers, half-filled with 10% neutral buffered formalin fixative, are used in histological applications for fixation and storage of tissue specimens.

  • Heavy-Duty Closure
  • Attached Handle
  • Heat Seal for 360 mL Containers
  • Click-Top Closure for 20 and 40 mL Containers
  • Paint Can Container with Attached Metal Lid Available

They are specifically designed for safe collection and transport of specimens

The 20 mL and 40 mL prefilled containers feature a click-top closure to ensure the user that the seal is fully engaged. Our 360 mL prefilled container features a heat seal to help prevent leaking during shipping.

Containers sized 20 mL through 120 mL are in convenient packs that allow for easy distribution to other labs and doctors offices. The packs are made of chipboard material, which is safe for operating room use. The 2.27 and 4.73 liter containers are made from high-density polyethylene. Each container features a heavy-duty screw top closure and an attached handle. We also offer a 3840 mL metal paint can container with attached metal lid.

Certificaciones: Sizes are 95 kPa compliant.

Información para pedidos: Minimum orders apply for the 0.6 gallon size (30 unit minimum) and the 1.25 gallon size (16 unit minimum).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR