VWR® Talon® Stainless Steel Fume Hood Lab-Frame Kits
Supplier: VWR, part of Avantor
VWR® Talon®
915748 915746 915745 915744
14224-406EA 1720.89 USD
14224-406 14224-404 14224-402 14224-400
VWR® Talon® Stainless Steel Fume Hood Lab-Frame Kits
Support Rods and Frames
Choose from four kits specifically designed to fit within laboratory fume hoods.
  • Stainless steel construction of rods
  • Nickel-plated zinc connectors and lab frame feet
  • Variety of kits designed to fit within laboratory fume hoods
The 1.5m (5') kit includes four 127 cm (50") horizontal rods, five 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 34 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 1.2m (4') kit includes four 96.5 cm (38") horizontal rods, four 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 30 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 1.8m (6') kit includes four 157.5 cm (62") horizontal rods, six 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 38 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 2.4m (8') kit includes four 218.5 cm (86") horizontal rods, eight 91.4 cm (36") vertical rods, three 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, ten 5.1 cm (2") rods, 45 lattice connectors, four rod-end connectors, and six Lab Frame feet.