VWR® Advanced and Standard Multi-Position Stirrers, 230 V (Export Only)
Supplier: VWR, part of Avantor
VWR®
80864008 80864000 80864012 80864004 80864015
12621-028EA 2464.76 USD
12621-028 12621-036 12621-048 12621-044 12621-020
VWR® Advanced and Standard Multi-Position Stirrers, 230 V (Export Only)
Stirrers Magnetic Stirrers, Multi-position
Microprocessor controlled
Synchronized and uniform mixing
Low-profile design
Best-in-Class, 5-year warranty
Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner.
  • Precise speed control
  • Cast metal base offers durability and added stability
  • Excellent low-speed stirring
Standard series offers an analog speed control knob with dial markings
Powerful magnetic stirrers with precise speed control offer repeatable performance. Synchronized operation ensures uniform mixing at each position. Ramping feature slowly brings sample to set speed to avoid splashing, improve magnetic coupling, and provide excellent low-speed stirring. Large top plate with position indicators is easy to clean and provides a white reflective surface. Low-profile design takes up less space and easily fits into fume hoods. Spill-resistant housing channels fluids away from internal components.
Advanced series offers an LED display for digital speed control, adjustable in 10rpm increments.
Certificaciones: RoHS certified. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Información para pedidos: Units include a 234 cm (92") Euro type plug. Units are also supplied with a 2.5 cm (1") PTFE-coated stir bar. Stirrers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.