Bochem Powder Coated Lab Support Jacks with Stainless Steel Lifts, BrandTech
Supplier: Bochem
B11015 B11090 B11040 B11080 B11020 B11030
12000-780EA 2780.38 USD
12000-780 12000-770 12000-772 12000-774 12000-776 12000-778
Bochem Powder Coated Lab Support Jacks with Stainless Steel Lifts, BrandTech
Laboratory Jacks
Improved–Jacks now feature thicker scissor mechanism supports for enhanced stability.
  • 304 stainless lift mechanisms for long service life
  • Epoxy-powder coated aluminum decks
Lift mechanism is constructed entirely of 304 stainless steel with POM rollers and phenolic knob for long service life in corrosive environments, such as fume hoods. All-roller design provides smooth stable height adjustment. Large (30x30cm and 40 x 40 cm) sizes are supplied with a ratcheting wrench for easy height adjustment. Jacks are manufactured according to DIN 12897.
Información para pedidos: Models B11080 and B11090 include ratchet to assist in adjustment.