VWR® General-Purpose Cleanroom Tape, Vinyl
Supplier: VWR, part of Avantor
VWR®
47B-15LG 47B-1RE 47B-2DG 47B-1PU 47B-1DB 47B-2RE 47B-3YE 47B-1TR 12YE-47B 47B-1YE 47B-2YE 47B-2LG 47B-1LB 47B-15WH 47B-34WH 47B-2BR 47B-1WH 47B-34YE 47B-12OR 47B-2WH 47B-1BL 47B-3WH 47B-12RE 47B-1DG 47B-2LB 47B-2DB 47B-1LG 47B-2OR 47B-34RE 47B-2TR 47B-12WH 47B-1OR 47B-15DB 47B-12DG 47B-2PU 47B-34DB
75779-612CS 491.54 USD
75779-612 11211-936 22221-075 11211-954 11211-938 22221-083 11211-978 11211-946 22221-044 11211-950 22221-086 22221-077 11211-940 11211-956 22221-050 68400-834 11211-914 22221-067 22221-047 11211-916 11211-948 22221-092 22221-040 11211-942 22221-073 22221-084 11211-944 22221-088 22221-062 22221-091 22221-028 11211-952 11211-962 89133-256 22221-081 22221-064
VWR® General-Purpose Cleanroom Tape, Vinyl
Tapes
VWR® General-Purpose Cleanroom Tape is ideal for use in cleanroom maintenance and support areas.
  • Multi-purpose vinyl tape
  • Color-coded for a variety of identification applications
  • 33m (36yd.) length wound on a plastic core
This multi-purpose vinyl tape is ideal for use in cleanroom maintenance and support areas. It is excellent for color coding, sealing, aisle marking, and batching. The matte finish can be written on with a cleanroom pen. Wound on a plastic core.
Información para pedidos: For additional colors and sizes, contact your VWR representative.