VWR® Construction Tape, Polyethylene
Supplier: VWR, part of Avantor
VWR®
CTP-2WH CTP-1WH
11211-912CS 445.37 USD
11211-912 11211-908
VWR® Construction Tape, Polyethylene
Tapes
This high-tack tape has rubber adhesive that easily bonds to plastic, concrete, and stainless steel.
  • Excellent conformity
  • Serrated edge for easy tearing by hand
  • Polyethylene fabrication
Tape conforms well and does not have the same “dead stretch” limitations common in other polyethylene tapes. Unique serrated edge allows an easy tear at any length, for environments where blades and dispensers are not practical.