VWR® Black Phenolic Bottle Caps
Supplier: VWR, part of Avantor
VWR®
292912 292974 292950 292943 292914 292963 292948 292970 292923 VWR89044-348 292983 292919 292935 292979 292954 292917 292962 292955 292972 VWR89044-374 292930 292957 292937 292932 292941 VWR89044-376 292928 VWR89044-284 VWR89044-344 292959 292984 292964 292931 292969 VWR89044-330 292920 292961 292939 292973 292953 292910 292976 292927 292978 292981 292980 292968 292965 VWR89044-342 292966 VWR89044-360 292925 292975 292949 292982 292956 VWR89044-368 292952 292913 292946
10862-426CS 214 USD
10862-426 10862-476 10862-432 10862-278 10862-474 10862-276 10862-436 10862-434 10862-478 10862-390 10862-270 10862-274 10862-472 10862-272 10862-418 10862-218 10862-222 10862-464 10862-388 10862-300 10862-462 10862-220 10862-468 10862-424 10862-224 10862-422 10862-302 10862-460 10862-448 10862-408 10862-454 10862-410 76329-638 10862-452 10862-298 10862-216 10862-412 10862-456 76329-632 10862-292 10862-290 76329-636 10862-296 10862-294 10862-438 10862-288 10862-442 10862-484 10862-286 10862-440 10862-446 10862-400 10862-280 76329-642 76329-640 10862-284 76329-646 10862-282 10862-480 76329-644
VWR® Black Phenolic Bottle Caps
Closures Bottle Closures
VWR® Black Phenolic Caps provide a wide range of chemical compatibility and temperature tolerance with an assortment of liners to choose from.
Pulp/Vinyl general purpose liners are commonly used for food, beverage, medical and chemical applications packed at less than 120 °F, including mild acids, alkalis, solvents, alcohols, oils and aqueous products; not recommended for active hydrocarbons or bleaches.
Solid PE liners offer good chemical resistance and low moisture vapor transmission rate. They are great for higher concentration acids and alkalis. Good for non-oil products filled at room temperature. Because this lining system is paperless, your samples will not be contaminated with fiber or paper particles from the closure.
Rubber lined caps are autoclavable. The liner is compliant with FDA 21 CFR 177.2600. Rubber articles intended for repeated use. The liner exhibits good resiliency and resistance to moisture vapor. The closures provide consistent adherence to close dimensional tolerance.
PE cone liners form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids to soaps and macro-invertebrates.
PTFE-faced foam liners resist attack from virtually all chemicals at room temperature and provides resilience for a tight seal with a low moisture transmission rate.
Envase: Packaged 12 bags of 12 caps per case for convenience.