VWR® Cell Culture Flasks Treated for Increased Cell Attachment, Sterile
Supplier: VWR, part of Avantor
CAF011850 CAF011600 CAF011250 CAF012850 CAF011025 CAF012250 CAF011050 CAF012050 CAF012025 CAF012600
10861-654CS 159.82 USD
10861-654 10861-644 10861-646 10861-672 10861-650 10861-640 10861-652 10861-642 10861-670 10861-648
VWR® Cell Culture Flasks Treated for Increased Cell Attachment, Sterile
Flasks Cell Culture Flasks
Increased Cell Attachment surface treatment can improve cell spreading and attachment
  • Available with 5 different growth areas
  • Increased Cell Attachment surface treated
  • Flask surface is flat and free from striation to maximize usable growth area
  • 2 different cap styles can be used in both open and closed systems
  • Innovative angled neck design offers good pipet and cell scraper access
  • Triangular top with wide base provides stability
  • Protruding ridge on the back side of the flask enables easy stacking
  • Special area near the neck for easy marking/writing
  • Engraved graduations on multiple sides
  • Strict integrity tested
  • Sterilized by electron beam irradiation
  • DNase- and RNase-free, and non-pyrogenic
The surface treatment is suitable for cells that may adhere poorly due to cell phenotype, stressful culture conditions, or those which normally require additional biological coatings for attachment.
Certificaciones: ISO13485 2003 and ISO9001 2008 certified.