BT Lab Systems Pipette Controller, G-Biosciences
Supplier: G-Biosciences
BT1514
10856-484EA 432.67 USD
10856-484
BT Lab Systems Pipette Controller, G-Biosciences
Pipets Pipet Controllers
A pipette controller that can fill 25 ml pipette in under 4 seconds.
  • Two different speed modes - High and Low along with gravity drain
  • The pump speed can be fine-tuned by varying finger pressure on operating knob to improve the sensitivity and for better control with low volumes
  • Cadmium Free environment friendly NiMH battery
  • Batteries can be changed very easily by user Intelligent charger prevents over charging and heating of batteries, with low battery indicator
  • For 0.1 to 200 ml plastic or glass pipettes
  • 8 hours continuous use 2 to 3 hour charge to full charge
  • Replaceable hydrophobic 0.45 µm PTFE filter and safety check valve
Pipette controller with double safety valves and filters to provide protection against fluid penetration.