VWR® Double-Sided Tape
Supplier: VWR, part of Avantor
VWDCL-12TRH VWDCL-1TRH VWDCL-12TRL VWDCL-1TRL
10853-864CS 2682.93 USD
10853-864 10853-858 10853-862 10853-860
VWR® Double-Sided Tape
Tapes
Transparent double coated polyester film available in Hi and Lo Tack with liner.
  • Double-coated
  • Polyester fabrication
Hi Tack features a permanent solvent acrylic adhesive—easily bonds dissimilar surfaces.
Lo Tack features a permanent adhesive on one side and removable on the other—ideal for peel and re-seal applications.