BRAND® Four Sided Clear Disposable Cuvettes for Fluorescence Applications, BrandTech® Scientific
Supplier: BrandTech
Brand®
759125 759030 759128 759035
10853-040PK 138.92 USD
10853-040 10853-044 10853-042 10853-038
BRAND® Four Sided Clear Disposable Cuvettes for Fluorescence Applications, BrandTech® Scientific
Cuvettes Spectrophotometer Cuvettes
Premium quality cuvettes, including a new UV-transparent plastic with ultra-low auto fluorescence.
  • Manufactured under cleanroom conditions
  • Polystyrene and UV-Transparent versions
BRAND® Fluorescence cuvettes are molded in one of the world's largest cleanrooms for life science plastics, without human contact for added cleanliness
It is used for routine, visible light applications, and for highest signal/noise ratios. They feature a 10 mm light path to fit all standard fluorimeters.
Cuvettes are mold-cavity sorted for the highest reproducibility.