VWR® Striped Safety Tape
Supplier: VWR, part of Avantor
VWSST-2REWH VWSST-2BLYE VWSST-2BLWH
10841-500CS 841.53 USD
10841-500 10841-232 10841-234
VWR® Striped Safety Tape
Tapes
Striped pressure sensitive tape for delineating and color coding specific equipment and areas.
  • Promotes plant safety
  • Can be used for marking emergency exits or safety equipment
This laminated vinyl tape is wound on a plastic core, is scuff resistant and holds up well to solvents, humidity and oil. Available in black/yellow, blue/white and red/white, SST can be used for marking fire equipment/exits, hazardous areas and first aid points within cleanrooms and controlled environments.