VWR® Plus Series Refrigerator and Freezer Combination Units
Supplier: VWR, part of Avantor
VWR®
HCRFC9SS SCRFC16A HCRFC9 HCRFC12G HCRFC12S
10819-794EA 6035.4 USD
10819-794 10819-926 10819-922 10819-924 10819-930
VWR® Plus Series Refrigerator and Freezer Combination Units
Refrigerators
These VWR® Plus Series refrigerator and freezer combo units are designed to provide laboratory-grade cooling and freezing capabilities in one streamline unit.
  • Microprocessor temperature controller
  • Two probe access ports (⅜"); one, refrigerator; one, freezer
  • Remote alarm contacts
  • Keyed door locks
  • Audible and visual high and low temperature alarms
  • HFC-free refrigerant
These refrigerator and freezer combo units are designed to deliver cooling results with economy of resources and space. Models regulate stored products through a series of temperature alarms for greater temperature control.
Units house shelves for product storage and come equipped with keyed door locks for product safety. All units feature two probe access ports, one for each compartment. The refrigerator and freezer combo units use natural, HFC-free refrigerant for economic efficiency and environmental cleanliness.
Certificaciones: UL/C-UL listed.