VWR® Signature™ Ergonomic High Performance Multichannel Pipettor
Supplier: VWR, part of Avantor
VWR® Signature™
P4308-300VWR 30709222 30709218 P08555-VE200 P4312-10VWR P4312-200VWR P4308-10VWR P4308-200VWR P08555-VE010 P4308-50VWR P12555-VE300 P4312-300VWR P4312-50VWR
89079-950EA 853.81 USD
89079-950 89079-952 89079-944 89079-954 89079-946 10002-842 89079-956 10813-582 10813-586 89079-948 10002-838 10002-848 89079-958
VWR® Signature™ Ergonomic High Performance Multichannel Pipettor
Pipets Micropipets
VWR® Signature™ Ergonomic high-performance Multichannel Pipettors provide superior accuracy and precision for repeatably reliable results.
  • Superior accuracy and precision for critical applications
  • Updated and improved ergonomic design
  • Volumes from 0.5 to 300 µl
  • Fully autoclavable
Innovative ergonomic features allow for consistently comfortable pipetting, even throughout prolonged use.
These lightweight, durable pipettors are fully autoclavable and supplied with a tool for in-lab calibration. Stand sold separately.
Designed to handle multiple samples simultaneously, pipettors are available in 8- or 12-channel formats. Upgraded ergonomic features on the multichannel models include reduced plunger and volume adjustment force to significantly reduce the strain associated with repetitive pipetting procedures. An extended, curved finger rest allows the pipettor to balance securely upon the index finger of the user.
Volume setting is continuously adjustable via the micrometer wheel or dispenser button, and can be locked to prevent accidental changes while in use. This also allows a variety of different volumes to be dispensed through a single pipettor, with no limitations on designated increments.
Tapered cones provide a universal tip fit and the adjustable tip ejector accommodates a variety of tips. An individual tip cone suspension system reduces the force necessary to load tips, as well as the force necessary to eject tips.