VWR® Tin Specimen Box
Supplier: VWR, part of Avantor
VWR®
11647-219 11647-220 11647-223 11647-222 11647-221
10806-398EA 169.44 USD
10806-398 10806-400 10806-394 10806-396 10806-402
VWR® Tin Specimen Box
Sample Containers General Purpose Containers
VWR® Tin Specimen Boxes are manufactured via an ASTM standard (A623-11) Tin-Milling process
  • Passivated
  • Round, seamless tin construction
  • Low-form, flat bottom design
  • Supplied with label on lid
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Boxes are excellent for sample collection and storage.