Puritan® Double Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
821-WCDBL 861-WCDBL 861-WCDBL 826-WCDBL
10806-004CS 414.07 USD
10806-004 89133-814 89133-810 CA10806-004
Puritan® Double Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Swabs Swabs for Sampling
For precision applications such as jewelry manufacturing or art restoration
Double-ended cotton head for dual use; one tapered end and one regular end. Rigid wood handle. Nonsterile. Made in USA.
Información para pedidos: Packaged 500 per box (packed as 5 bags of 100), 10 boxes per case.