Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
806-WCL 806-WCL 806-WC 806-WC
10806-001CS 347.9 USD
CA10806-001 10806-001 10806-000 CA10806-000L
Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Swabs Swabs for Sampling
These Puritan® swabs are ideal for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens.
Swabs with adhesive incorporate a nonsterile cotton tip secured by adhesive to the wooden handle. Swabs are also available without adhesive, for use where contamination of critical surfaces is a concern. Swabs are autoclavable.
They are also useful for cleaning delicate laboratory instruments.
Información para pedidos: Packaged in bags of 100 swabs, 10 bags to a pack, 10 packs to a case.