Pur-Wraps® Swab, Cotton Tip, Sterile or Nonsterile, Puritan Medical Products
Supplier: Puritan Medical Products
Pur-Wraps®
25-8061PC 803-PC 803-PC 25-8061PC 806-PC 25-8062PC 25-8062PC 806-PC
10805-148CS 820.99 USD
10805-148 CA10805-144 10805-144 CA10805-140 10806-050 10805-140 CA10806-050 CA10805-148
Pur-Wraps® Swab, Cotton Tip, Sterile or Nonsterile, Puritan Medical Products
Swabs Swabs for Sampling
This absorbent cotton tipped applicator is produced with high-quality USP grade cotton, which is carded and coiled onsite to maintain consistency.
It is bonded securely to a semi-flexible polystyrene handle by an aqueous-based adhesive. Available either nonsterile or sterile.