VWR® Specimen Containers with Dual Click
Supplier: VWR, part of Avantor
VWR®
145530 242220 242022 244020 242520 243220 243920 244220 242020 245930 244021 244920 245230 142020 242920 143220 242620 142620 243420 142220 144220 242420 145230 144620 244520 245530 243020 243021 245030 145030 242021
10803-990CS 349.04 USD
10803-990 10804-034 10804-012 10804-036 10804-018 10803-968 10803-988 10804-042 10803-960 10803-964 10803-986 10803-962 10803-984 10804-000 10804-024 10804-046 10804-004 10804-048 10803-978 10803-956 10804-050 10804-052 10803-972 10803-994 10803-950 10803-992 10803-976 10803-998 10803-952 10803-996 10803-974
VWR® Specimen Containers with Dual Click
Sample Containers General Purpose Containers
The VWR® Dual Click line of specimen containers are manufactured from virgin medical grade propylene, and the Dual Click design features two audible and tactile “clicks” to let the user know the lid is properly secured.
  • Virgin medical grade propylene construction
  • Audible and tactile clicks ensure a secure seal
  • Available in sterile and nonsterile formats
Dual Click closures ensure safe specimen transport
Sterile Dual Click specimen containers are sterilized using E-Beam sterilization. Individually wrapped sterile Dual Click containers are also available for the ultimate in sterility assurance. Certificates of sterility are available for all sterile products. Container 10804-046 also includes a sodium thiosulfate tablet for water testing procedures.
Once the specimen is collected, a simple turn of the closure will give the user assurance that the seal is fully engaged with a “click” sound. The second “click” ensures 95 kPa compliance for pneumatic tube transportation. All containers are manufactured in the USA.