Motic BA310 Elite Upright Compound Microscope
Supplier: Motic
1100100402432 1100100402632 1100100402432 1101001100401 1100100402631 1100100402642 1100100402641 1100100402442 1101001100401
10768-194EA 1857.45 USD
10768-194 10767-582 CA10769-642 10768-196 10767-580 10767-578 CA89168-482 89168-482 10768-192
Motic BA310 Elite Upright Compound Microscope
Microscopes
Motic’s BA310 Elite Series is built for the performance demands of colleges, universities, and medical school markets, setting a new price-performance standard for optical quality
The BA310 Elite model introduces superior improvements in both optical and mechanical performance. The rackless stage concept without a prominent gear rack permits even more convenient x/y movement and effortless positioning of the stage without gear rack interference. Ergonomic drop-down coaxial controls enable convenient 'eyes down' operation for uninterrupted sample viewing. The included CCIS EC Plan Achromatic lenses are multilayer-coated for improved color fidelity and image enhancement, giving superior image reproduction.
The improved BA310 Elite series introduces innovative features like second-generation EC Plan optics, a new rackless stage movement, and an advanced modular illumination design offering halogen/LED interchangeability. The BA310 Elite series brings versatility and function to a wide range of education and professional applications.
Información para pedidos: Microscope is supplied with micrometer reticles, phase contrast accessories, polarizing accessories, and Darkfield accessories. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative. All Motic products come with a one-year electrical and five-year mechanical warranty.