SMZ-171 Stereo Microscope, Motic
Supplier: Motic
1101001402941 1101001402711 1101001402931 1100200600762 1101001402951 1100200600767 1101001402741 1101001402961 1101001402721 1100200600753 1100200600767 1100200600758 1101001402731
10767-556EA 224.68 USD
10767-556 10767-534 10767-546 10767-554 10767-544 10767-538 CA10769-604 10767-536 10767-548 10767-552 10767-542 10767-550 10767-540
SMZ-171 Stereo Microscope, Motic
Microscopes
Flexible stereo microscopes for advanced research applications in biomedicine and material science. The 6.7:1 Greenough zoom delivers a standard magnification of 7.5x - 50x with 110 mm working distance in a FOV23mm.
Información de suministro: Supplied with the following: Stands, Stage Accessories, Micrometer Reticles, Additional Illumination, Polarizing Equipment, Darkfield Equipment, Barlow Lenses. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative.