VWR® Disposable, Graduated, Plastic Transfer Pipet
Supplier: VWR, part of Avantor
VWR®
201C.US 200CS20 200CS01 200C.US 218C.US 212C.US
10754-264BX 272.79 USD
10754-264 10755-252 10754-268 10754-266 10754-272 10754-270
VWR® Disposable, Graduated, Plastic Transfer Pipet
Pipets Pasteur/Transfer Pipets
VWR® Disposable Transfer Pipets are designed to safely transfer and dispense liquids in all types of laboratories.
  • Pipets are packaged using medical grade paper and plastic film
  • Non-toxic pipets can be safely incinerated or heat-sealed and frozen at temperatures as low as –70 °C (–94 °F)
VWR® pipets are produced using an optimal blend of low density polyethylene, allowing for highly controlled liquid dispensing.