VWR® C-Leg Bench Frame with Top for Double Bay Uprights
Supplier: VWR, part of Avantor
14-C100VWR499 14-C100VWR495 14-C103VWR496 14-C101VWR492 14-C100VWR493 14-C103VWR495 14-C101VWR496 14-C100VWR498 14-C100VWR497 14-C100VWR496 14-C103VWR493 14-C101VWR499 14-C101VWR493 14-C103VWR497 14-C103VWR492 14-C101VWR497 14-C101VWR494 14-C101VWR495 14-C101VWR498 14-C103VWR499 14-C103VWR494 14-C103VWR498 14-C100VWR494 14-C100VWR492
10682-708EA 4737.8 USD
10682-708 10682-706 10682-674 10682-696 10682-684 10682-694 10682-672 10682-682 10682-670 10682-692 10682-680 10682-690 10682-704 10682-714 10682-702 10682-712 10682-668 10682-678 10682-700 10682-710 10682-688 10682-676 10682-698 10682-686
VWR® C-Leg Bench Frame with Top for Double Bay Uprights
Furniture Workbenches
These VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads (static) up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution. Frame color is light neutral.
Información para pedidos: Configurations include frame (pair of legs, rear case, and front rail) and top for double bay uprights (uprights not included). Top includes black phenolic or black laminate options.