VWR® Graduated Cylinders, Calibrated To Deliver, Class B
Supplier: VWR, part of Avantor
138.288.08A 138.288.03A 138.288.06A 138.288.02A 138.288.07A 138.288.05A 138.288.01A 138.288.04A
10546-062CS 325.27 USD
10546-062 10546-060 10545-970 10546-058 10546-056 10546-054 10545-968 10546-052
VWR® Graduated Cylinders, Calibrated To Deliver, Class B
Cylinders Measuring Cylinders
Cylinder sizes from 25mL to 2000mL feature a hexagonal base and yellow bumper.
  • Borosilicate glass fabrication
  • Calibrated at 20°C to deliver
“TD” appears on each cylinder and indicates the cylinder is calibrated to deliver
Made from ASTM E438 Type 1 Class A borosilicate 3.3 Glass. As per ASTM Specification E1272 Class B requirements.
Durable white color scale printing for long life in laboratory environment.