VWR® Erlenmeyer Flasks, Narrow Mouth
Supplier: VWR, part of Avantor
VWR®
231.402.01 231.435.07 VWRI75870-512 231.435.05 231.402.04A 231.402.02 231.402.03A 231.402.04 231.435.08A 231.402.09 231.435.06
10536-676CS 353.13 USD
10536-676 10545-844 75870-512 10536-926 10536-924 10536-678 10536-912 10536-914 10545-846 10545-848 10545-842
VWR® Erlenmeyer Flasks, Narrow Mouth
Flasks Erlenmeyer Flasks
Flasks are durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.
  • Borosilicate glass fabrication
  • Durable design
Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.